Order Entry
United States
ContactUsLinkComponent
Date of Inspection Labels, Labelmaster
Date of Inspection Labels, Labelmaster
Catalog # 101376-330
Supplier:  Label Master
Date of Inspection Labels, Labelmaster
Catalog # 101376-330
Supplier:  Label Master
Supplier Number:  FIL-91

Specifications

  • Description:
    Inspection labels
  • Dimensions:
    152.4×152.4 mm (6×6")
  • Material:
    Permanent vinyl
  • Thickness:
    11.2 mil.
  • Cat. No.:
    101376-330
  • Legend:
    Date Of Inspection
  • Supplier no.:
    FIL-91

Specifications

About this item

Manage fleet inspection status dates with Date of Inspection labels.

  • PVC-free vinyl labels

Designed to highlight the previous inspection date and the next scheduled inspection date, each label can be signed and dated by the supervisor or mechanic responsible for the inspection.