Arrows Up/This End Up Labels, Labelmaster
101378-200
- Description:Arrows up labels
- Dimensions:101.6×228.6 mm (4×9")
- Material:Semi-gloss paper
- Thickness:5.85 mil.
- Cat. No.:101378-200
- Legend:This Side Up
- Supplier no.:L400
Comply with air shipment regulations with Arrows Up/This End Up labels.
- Prionted on paper
- Outstanding chemical and abrasion resistance
- Strong adhesion to corrugated boxes
Labels comply with the requirements in International Air Transport and ICAO for marking the package orientation on a hazardous materials package.