ContactUsLinkComponent
KIMBLE® Microscale Tissue Grinder Kit with Grinder and Pestle Varieties, DWK Life Sciences
KIMBLE® Microscale Tissue Grinder Kit with Grinder and Pestle Varieties, DWK Life Sciences
Catalog # KT884900-0000
undefined
KIMBLE® Microscale Tissue Grinder Kit with Grinder and Pestle Varieties, DWK Life Sciences
Catalog # KT884900-0000
Supplier Number:  884900-0000
Specifications
  • Description:
    Tissue grinder kit
  • Environmentally Preferable:
  • Cat. No.:
    KT884900-0000
  • Supplier no.:
    884900-0000
Specifications
About this item
Kit contains a variety of the finest microscale tissue grinders available. It includes ground glass, smooth glass and PTFE grinders suitable for tough connective tissue and soft tissue such as brain or liver.
  • Working with micro quantities reduces heat build-up and yields greater enzyme activity
  • Samples can be handled with minimal loss; transfers can often be eliminated
  • Grinders are also available individually
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Packaging: Items are packaged in a convenient high-density polyethylene case with die cut foam inserts that protect grinders in transit and in storage.