Order Entry
United States
ContactUsLinkComponent
Fixed-Speed Vortex Mixer
Fixed-Speed Vortex Mixer
Catalog # 470319-914
undefined
Fixed-Speed Vortex Mixer
Catalog # 470319-914
Supplier Number:  8031211000

Specifications

  • Description:
    Fixed speed vortex mixer
  • Dimensions:
    5×5×3"
  • Cat. No.:
    470319-914
  • Electrical:
    100/220 V, 50/60 Hz
  • Orbit:
    4.8 mm
  • Speed:
    3000 rpm

Specifications

About this item

This small vortexer conserves bench-space but is powerful enough for most applications.

  • Fixed Speed
  • Touch-activated operation
  • DC Brushless motor provides stable performance