KIMBLE® ULTRA-WARE® Debubbler, DWK Life Sciences
Catalog # KT953890-0000
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:ULTRA-WARE® debubbler
- Environmentally Preferable:
- Cat. No.:KT953890-0000
- Supplier no.:953890-0000
Specifications
About this item
Unit consists of a glass chamber with inlet and outlet ports and a PTFE - lined phenolic cap.
- Installed before the pump or mixer, the debubbler captures bubbles that are released by unscrewing the cap
- Cap GPI Thread: 13-425
- Port threads 14"-28
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Unions are not included.