Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Water
Water, Oakton®, deionized Water Rinse Solution, Pouches
Water, Oakton®, deionized Water Rinse Solution, Pouches
Water, Oakton®, deionized Water Rinse Solution, Pouches
76371-714
:
Antyila Scientific
:
Water, Oakton®, deionized Water Rinse Solution, Pouches
76371-714
:
Antyila Scientific
:
35653-00
:
Pack type:
Pouch
Size:
20×20 ml
Cat. No.:
76371-714
:
:
18.02 g/mol
:
7732-18-5
:
H₂O
:
MFCD00011332