ContactUsLinkComponent
KIMBLE® KONTES® Cold Finger, Condenser, DWK Life Sciences
KIMBLE® KONTES® Cold Finger, Condenser, DWK Life Sciences
Catalog # KT746065-0000
undefined
KIMBLE® KONTES® Cold Finger, Condenser, DWK Life Sciences
Catalog # KT746065-0000
Supplier Number:  746065-0000
Specifications
  • [ST] joint:
    14/10
  • Cat. No.:
    KT746065-0000
  • Fits:
    ¼"
  • Supplier no.:
    746065-0000
Specifications
About this item
Drip tip on the bottom directs drops into the collar of Hickman distillation heads.
  • Micro-adjustable
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
This cold finger condenser is used as an accessory for microscale Standard Taper 14/10 threaded kits.