ContactUsLinkComponent
Hickman Distillation Head, with Side Arm, DWK Life Sciences
Catalog # KT746080-0000
undefined
Hickman Distillation Head, with Side Arm, DWK Life Sciences
Catalog # KT746080-0000
Supplier Number:  746080-0000
Specifications
  • [ST] joint:
    14/10
  • Description:
    Hickman distillation head
  • Cat. No.:
    KT746080-0000
  • Supplier no.:
    746080-0000
Specifications
About this item
Glassware manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
  • Ideal for use to 200 °C