KIMAX® Straight-Bore Stopcock, without Plug, DWK LIFESCIENCES
Catalog # 59140-088
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Stopcock, straight bore, without plug
- Cat. No.:59140-088
- Stopcock size:4 mm
- Supplier no.:41004G-4
Specifications
About this item
Straight-bore, sandblasted stopcock with a 1:5 taper.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.