CD-Chex CD117® Plus Immunophenotyping Controls, Streck
Supplier: Streck Laboratories
Certificates
About this item
Flow cytometry controls for CD117, CD25 and CD71.
- Exhibits abnormal peripheral blood characteristics
- Positive procedural control
- Assay values for abnormal and total leukocytes
These controls have a 90-day closed-vial stability and a 30-day open-vial stability.
Frequently Bought Together