Filling nozzle clamp
Catalog # EM9.67107.0001
Supplier: MilliporeSigma
Specifications
- Description:Filling nozzle clamp with wall attachment
- Environmentally Preferable:
- Cat. No.:EM9.67107.0001
- Supplier no.:9.67107.0001
Specifications
About this item
Stainless steel clamp for filling nozzles, with drip-free attachment to walls.
For individual solvent withdrawal installations that end with a flexible filling nozzle inside a fume hood.