



n-Hexane ≥99.0% (by GC), LiChrosolv®, hypergrade for LC-MS, Supelco®
Catalog # EM1.03701.2500
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:EM1.03701.2500
Specifications
Specification Test Results
- Purity (GC):≥99.0%
- Identity (IR):Conforms
- Evaporation Residue:≤2.0 mg/l
- Water:≤0.005%
- Acidity:≤0.0002 meq/g
- Alkalinity:≤0.0002 meq/g
- Al (Aluminium):≤10 ppb
- Ca (Calcium):≤10 ppb
- Fe (Iron):≤10 ppb
- K (Potassium):≤5 ppb
- Mg (Magnesium):≤10 ppb
- Na (Sodium):≤10 ppb
- Transmission (at 210 nm):≥60%
- Transmission (at 225 nm):≥ 90%
- Transmission (from 245 nm):≥98%
- Suitable for LC-MS; Intensity of Background Mass Peak Based on Reserpine (APCI/ESI Positive):≤2 ppb
- Suitable for LC-MS; Intensity of Background Mass Peak Based on Reserpine (APCI/ESI Negative):≤20 ppb
Specification Test Results
About this item
:
:86.18 g/mol
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520