Order Entry
United States
ContactUsLinkComponent
n-Heptane ≥99.3% (by GC), LiChrosolv®, hypergrade for LC-MS, Supelco®
Flammable
Health
Irritant
Environment
n-Heptane ≥99.3% (by GC), LiChrosolv®, hypergrade for LC-MS, Supelco®
Catalog # EM1.03654.2500
Supplier:  MilliporeSigma
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
n-Heptane ≥99.3% (by GC), LiChrosolv®, hypergrade for LC-MS, Supelco®
Catalog # EM1.03654.2500
Supplier:  MilliporeSigma
Supplier Number:  1.03654.2500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    2.5 L
  • Cat. No.:
    EM1.03654.2500

Specifications

Specification Test Results

  • Purity (GC):
    ≥99.3%
  • Identity (IR):
    Conforms
  • Evaporation Residue:
    ≤2.0 mg/l
  • Water:
    ≤0.005%
  • Acidity:
    ≤0.0002 meq/g
  • Alkalinity:
    ≤0.0002 meq/g
  • Al (Aluminium):
    ≤ 10 ppb
  • Ca (Calcium):
    ≤10 ppb
  • Fe (Iron):
    ≤10 ppb
  • K (Potassium):
    ≤5 ppb
  • Mg (Magnesium):
    ≤10 ppb
  • Na (Sodium):
    ≤10 ppb
  • Transmission (at 210 nm):
    ≥50%
  • Transmission (at 220 nm):
    ≥80%
  • Transmission (from 245 nm):
    ≥98%
  • Suitable for LC-MS; Intensity of Background Mass Peak Based on Reserpine (APCI/ESI Positive):
    ≤2 ppb
  • Suitable for LC-MS; Intensity of Background Mass Peak Based on Reserpine (APCI/ESI Negative):
    ≤20 ppb

Specification Test Results

About this item

 :
 :100.20 g/mol
Boiling Pt:97...98 °C (1013 hPa)
Melting Pt:–90.5 °C
Density:0.68 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544