

Ethyl acetate ≥99.8% (by GC), LiChrosolv®, hypergrade for LC-MS, Supelco®
EM1.03649.2500
:
- Pack type:Glass bottle
- Size:2.5 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 solvent selection guide recommended by default and after discussion
- Cat. No.:EM1.03649.2500
- Purity (GC):≥99.8%
- Identity (IR):conforms
- Evaporation Residue:≤2.0 mg/l
- Water:≤0.05%
- Acidity:≤0.0002 meq/g
- Alkalinity:≤0.0002 meq/g
- Al (Aluminium):≤10 ppb
- Ca (Calcium):≤10 ppb
- Fe (Iron):≤10 ppb
- K (Potassium):≤5 ppb
- Mg (Magnesium):≤10 ppb
- Na (Sodium):≤50 ppb
- Transmission (at 260 nm):≥50%
- Transmission (at 265 nm):≥80%
- Transmission (from 270 nm):≥98%
- Suitable For LC-MS; Intensity Of Background Mass Peak Based On Reserpine (APCI/ESI Positive):≤2 ppb
- Suitable For LC-MS; Intensity Of Background Mass Peak Based On Reserpine (APCI/ESI Negative):≤20 ppb
:
:88.11 g/mol
:77 °C (1013 hPa)
:–83 °C
:0.90 g/cm³ (20 °C)
:–4 °C
:Ambient
:205-500-4
:1173
:3,II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS07
:141-78-6
:CH₃COOC₂H₅
:MFCD00009171