Flame Resistant Deluxe Coveralls, MCR Safety
These coveralls are manufactured from a comfortable 88% cotton/12% nylon twill material.
- Max Comfort™ 9 oz., 88% cotton/12% nylon FR twill fabric
- Two patch pockets on chest with snap closure
- Two front set-in hip pockets
- Side vent pass throughs
- Two back patch pockets with concealed snap closures
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in FR gear made with Max Comfort™ fabrics. Garments feature FR reflective material for high visibility in low light areas.
: ASTM F1506-08 electric arc protection standard; NFPA 70E-2012 hazard/risk category HRC 2; ATPV ASTM 1959 11.7 cal/cm², HRC 2; CAN/CGSB155.20-2000 CAN/CGSB155.22-97, OSHA 29 CFR 1910.269; CAN/CSA Z96-09 CLASS 1 / LEVEL FR. Meets the NFPA 2112-2018 standard for flame resistant garments and NFPA 70E standard for electrical safety in the workplace.
- Catalog No:
- 76341-468
- 76341-466
- 76341-870
- Garment application:
- Industrial
- Industrial
- Industrial
- Risk area:
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- HiVis, Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Chest long
- Chest long
- Chest long
- Closure type:
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- Fabric:
- Max Comfort™, 88% cotton/12% nylon FR twill fabric
- Max Comfort™, 88% cotton/12% nylon FR twill fabric
- Max Comfort™, 88% cotton/12% nylon FR twill fabric
- Thickness:
- 9 oz.
- 9 oz.
- 9 oz.
Frequently Bought Together