Flame Resistant Contractor Coveralls, MCR Safety
These Coveralls are manufactured from a comfortable 100% cotton material.
- Max Comfort™ 7 oz., 100% cotton FR twill fabric
- Two-way concealed Nomex® FR brass break-away zipper
- One patch pocket on chest
- Side vent pass throughs
- Five-thread overlock stitching
- Two front pockets and two rear patch pockets on coverall pants
- Sewn with Nomex® FR thread
- One-piece, top stitched, lay flat collar
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in FR Gear made with Max Comfort™ fabrics.
: NFPA 2112 UL certified. ASTM F1506 electric arc protection. NFPA 70E hazard/risk Cat HRC2/ATPV ASTM 1959 9.6 cam/cm². Meets the NFPA 2112-2018 standard for flame resistant garments and NFPA 70E standard for electrical safety in the workplace.
- Catalog No:
- 76341-304
- Garment application:
- Industrial
- Risk area:
- Flame-retardant
- Closure type:
- With Zip Closure
- Fabric:
- 100% cotton FR twill
- Weight:
- 7 oz.
Frequently Bought Together