Flame Resistant Extreme Climate Insulated Bib Overalls with Wind/Vapor Barrier, MCR Safety
Extreme climate insulated overalls are manufactured from a modacrylic quilted lining and a wind/vapor barrier for warmth and protection.
- Max Comfort™ 9 oz., 88% cotton/12% nylon outer shell
- 11.5 oz. modacrylic insulation/3 oz. wind and vapor barrier
- Two front set-in hip pockets
- Vented legs with storm flap protection and snap closure
- Two rear pockets with concealed snap closures
- Patch pockets on chest
- Elasticized waist for comfort and security
- Sewn with FR aramid thread
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in FR gear made with Max Comfort™ fabrics.
: ASTM F1506-08 electric arc protection standard. NFPA 70E-2018 hazard/risk category, HRC3. ATPV ASTM 1959 38 cal/cm²; CAN/CGSB155.20-2000 CAN/CGSB155.22-97, OSHA 29 CFR 1910.269; CAN/CSA Z96-09 CLASS 1/LEVEL FR. Max Comfort™ material meets the NFPA 2112-2018 standard for flame resistant garments and NFPA 70E standard for electrical safety in the workplace.
- Catalog No:
- 76342-328
- 76341-588
- Garment application:
- Industrial, Fire protection, Oils, fats and fuels
- Industrial, Fire protection, Oils, fats and fuels
- Risk area:
- Flame-retardant
- Flame-retardant
- Gender:
- For Men
- For Men
- Sleeve or leg length:
- Long
- Long
- Fabric:
- 88% cotton/12% nylon
- 88% cotton/12% nylon
Frequently Bought Together