Flame Resistant Break-Away Safety Vests, Class 2, MCR Safety
Supplier: MCR Safety
Certificates
About this item
ANSI rated safety vests are specifically designed for high visibility needs.
- FR solid modacrylic/aramid blend fabric, fluorescent lime finish
- 2" silver FR reflective stripes
- 5-point break-away vest with hook and loop closures
- No pockets
This flame-resistant safety vest is made with GlenGuard® solid lime modacrylic/aramid blended fabric with FR reflective stripes. Vests are equipped with durable gray binding and an FR hook and loop front closure.
Certifications: ANSI/ISEA 107-2015 Class 2, Type R compliant; Conforms to NFPA 70E, HRC1, arc rating 5.4 cal/cm², meets ASTM F1506.
Specifications
- Catalog No:
- 76340-788
- 76340-712
- Garment application:
- Fire protection, General Purpose
- Fire protection, General Purpose
- Risk area:
- HiVis
- HiVis
- Gender:
- Unisex
- Unisex
- Sleeve or leg length:
- Short
- Short
- Closure type:
- With Velcro
- With Velcro
- Fabric:
- Solid lime modacrylic/aramid
- Solid lime modacrylic/aramid