Flame Resistant Extreme Climate Insulated Coveralls, MCR Safety
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in FR gear made with Max Comfort™ fabrics.
- Two patch pockets on chest with snap closure
- Two front set-in hip pockets and two back patch pockets with concealed snap closures
- Modacrylic quilted lining with wind and vapor barrier
- Elasticized waist for comfort and security
- Sewn with FR Aramid thread
- All metal in garment is protected to avoid exposure to skin
- FR yellow-silver-yellow reflective tape: arms, legs, complete stripes on front, X on back with complete belt
These coveralls are manufactured from a comfortable 88% cotton/12% nylon twill material. They also feature a modacrylic quilted lining for protection and warmth, and a wind and vapor barrier for outstanding comfort.
: Material that meets the NFPA 2112-2018 standard for flame resistant garments and NFPA 70E standard for electrical safety in the workplace. ASTM F1506-08 electric arc protection standard. NFPA 70E-2012 hazard/risk category HRC3. ASTM 1959, ATPV 38 cal/cm²; CAN/CGSB155.20-2000; CAN/CGSB155.22-97, OSHA 29 CFR 1910.269; CAN/CSA Z96-09 CLASS 1/LEVEL FR.
- Catalog No:
- 76342-596
- 76342-594
- 76342-590
- 76342-588
- 76342-390
- 76342-388
- Garment application:
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Risk area:
- Anti-static, Flame-retardant
- Anti-static, Flame-retardant
- Anti-static, Flame-retardant
- Anti-static, Flame-retardant
- Anti-static, Flame-retardant
- Anti-static, Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- With Zip Closure
- Fabric:
- 88% cotton and 12% nylon
- 88% cotton and 12% nylon
- 88% cotton and 12% nylon
- 88% cotton and 12% nylon
- 88% cotton and 12% nylon
- 88% cotton and 12% nylon
Frequently Bought Together