Plain Glass Microscope Slides
Catalog # 470320-820
Supplier: United Scientific Supplies
Specifications
- Description:Plain slides with rounded corners
- Cat. No.:470320-820
Specifications
About this item
Pre-cleaned slides with rounded corners.
- Made of quality sheet glass, with ground edges
These slides are 75×25 mm, 1.0 to 1.2 mm thick, and are packaged in boxes of 72.