ContactUsLinkComponent
Difco™ Yeast Extract Glucose Chloramphenicol Agar, BD Life Sciences
Difco™ Yeast Extract Glucose Chloramphenicol Agar, BD Life Sciences
Catalog # 95062-859
Supplier:  BD
CAS Number:  
Difco™ Yeast Extract Glucose Chloramphenicol Agar, BD Life Sciences
Catalog # 95062-859
Supplier:  BD
Supplier Number:  219001
CAS Number:  
Specifications
  • Description:
    Difco™ Yeast extract glucose chloramphenicol agar
  • Size:
    500 g
  • Cat. No.:
    95062-859
  • Supplier no.:
    219001
Specifications
About this item
Yeast Extract Glucose Chloramphenicol Agar is a selective agar highly recommended for enumerating yeasts and molds in milk and milk products.
  • Beige, free-flowing and homogeneous
Certifications: Recommended by the International Dairy Federation 1,2.