BioClean™ Sterile Chemotherapy Prep Bench Mat, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
Sterile chemotherapy prep mats are highly absorbent.
- Three-layer construction
- Mesh upper layer for even distribution, highly absorbent middle layer, impermeable bottom layer
- Sterilised by gamma radiation
- Low-linting and latex-free
The BioClean™ Chemotherapy Prep Bench Mat enhances workbench safety for technicians, nurses, and pharmacists when handling or administering chemotherapy drugs. The prep mat is highly-absorbent, three-layered preparation mat consisting of two layers of polyethylene with a central layer of air-laid paper. The upper layer allows even distribution of liquid, the middle layer gives absorption properties, and the bottom layer provides defense against leakages.
Certifications: Manufactured in a facility holding ISO 9001:2008 certifications, Processed in an NEBB certified ISO Class 5 environment.
Caution: To be stored in a dry, cool place (up to 40 °C) away from direct sunlight and fluorescent light.
Frequently Bought Together