Order Entry
United States
ContactUsLinkComponent
WHEATON® Bottle Top Vacuum Filtration System, DWK Life Sciences
WHEATON® Bottle Top Vacuum Filtration System, DWK Life Sciences
Catalog # 10014-238
undefined
WHEATON® Bottle Top Vacuum Filtration System, DWK Life Sciences
Catalog # 10014-238
Supplier Number:  419520-47

Specifications

  • Pore size:
    40.00 - 60.00 µm
  • Description:
    Glass filtration complete unit, with fritted glass support and GL 45 thread connection
  • Cat. No.:
    10014-238
  • Membrane diameter:
    47 mm
  • Supplier no.:
    419520-47

Specifications

About this item

The system is designed to collect the filtrate directly into a standard laboratory bottle.

  • Designed to collect the filtrate directly into a standard laboratory bottle
  • Connection to the bottle is made with a polybutylene terephthalate (PBT) cap and a PTFE-faced silicone sealing ring
  • Recommended for general filtration

Glass components are manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.

Delivery: Includes a coarse porosity (40 to 60 µm) fritted glass support base, graduated funnel (300 ml), anodized aluminum clamp, and reservoir (1 L).