Linearity Vitamin D, AUDIT Microcontrols
About this item
Linearity LQ Vitamin D, Linearity FLQ Vitamin D for Ortho Vitros and Linearity LQ Vitamin D for Roche systems are assayed quality control materials consisting of five levels.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
1 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Linearity LQ Vitamin D, Linearity FLQ Vitamin D for Ortho Vitros and Linearity LQ Vitamin D for Roche systems are assayed quality control materials consisting of five levels.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Documents
Product Family Options
Product Information
- TypeAnalyteStabilitySizeAvailabilityPrice
- 76359-738Linearity FLQ Vitamin D for Ortho VitrosLinearity FLQ Vitamin D for Ortho VitrosFrozen liquidVitamin D7 days when stored at 2...8 °C5×1 mlLog in for availabilityKit of 5 (1ml)$382.39Supplier Number: K910M-5Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Linearity LQ Vitamin D for Roche SystemsLiquidVitamin D14 days when stored at 2...8 °C5×2 mlDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Linearity LQ Vitamin DLiquidVitamin D14 days when stored at 2...8 °C5×2 mlDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.