Linearity TIBC, AUDIT Microcontrols
Catalog # 76359-734
Supplier: Audit Microcontrols
CAS Number:
Specifications
- Description:Linearity DROP LQ TIBC for Ortho Vitros
- Size:5×2 ml
- Cat. No.:76359-734
- Stability:5 days when stored at 2...8 °C
- Analyte:TIBC
- Type:Liquid
Specifications
About this item
Linearity DROP LQ TIBC is assayed quality control material consisting of five levels of bovine based serum albumin.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.