Linearity Theraputic Drug Monitoring, AUDIT Microcontrols
Supplier: Audit Microcontrols
Certificates
About this item
Universal TDM Linearity and Siemens Centaur TDM Linearity are assayed quality control materials consisting of five levels of human serum.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Frequently Bought Together