Order Entry
United States
ContactUsLinkComponent
Linearity Protein, AUDIT Microcontrols
Linearity Protein, AUDIT Microcontrols
Catalog # 97068-884
CAS Number:  
undefined
Linearity Protein, AUDIT Microcontrols
Catalog # 97068-884
Supplier Number:  K702M-5
CAS Number:  

Specifications

  • Description:
    Linearity LQ protein
  • Size:
    5×2 ml
  • Cat. No.:
    97068-884
  • Stability:
    24 hours when stored at 2...8 °C
  • Analyte:
    Alpha1-Antitrypsin, Complement C3, Complement C4, Immunoglobulin G (IgG), Immunoglobulin M (IgM), Immunoglobulin A (IgA), Protein, Total, Transferrin (TRF)
  • Type:
    Liquid
  • Supplier no.:
    K702M-5

Specifications

About this item

Protein linearity is assayed quality control material consisting of five levels of human based serum.

Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.