Order Entry
United States
ContactUsLinkComponent
Linearity Microalbumin/Microprotein, AUDIT Microcontrols
Linearity Microalbumin/Microprotein, AUDIT Microcontrols
Catalog # 97068-910
CAS Number:  
undefined
Linearity Microalbumin/Microprotein, AUDIT Microcontrols
Catalog # 97068-910
Supplier Number:  K722M-5
CAS Number:  

Specifications

  • Description:
    Linearity LQ microalbumin/microprotein
  • Size:
    5×2 ml
  • Cat. No.:
    97068-910
  • Stability:
    7 days when stored at 2...8 °C
  • Analyte:
    Microalbumin, microprotein
  • Type:
    Liquid
  • Supplier no.:
    K722M-5

Specifications

About this item

Microalbumin/Microprotein linearity is assayed quality control material consisting of five levels of human based serum.

Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.