Linearity hs-CRP, AUDIT Microcontrols
Catalog # 97068-890
Supplier: Audit Microcontrols
CAS Number:
Specifications
- Description:Linearity LQ hs-CRP
- Size:5×1 ml
- Cat. No.:97068-890
- Stability:10 days when stored at 2...8 °C
- Analyte:hs-CRP
- Type:Liquid
- Supplier no.:K705M-5
Specifications
About this item
Linearity LQ hs-CRP is assayed quality control material consisting of five levels of human based serum.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.