Linearity Glycohemoglobin A1c, AUDIT Microcontrols
Supplier: Audit Microcontrols
Certificates
About this item
Glycohemoglobin A1c linearity is assayed quality control material consisting of five levels of human blood. Linearity FLQ HbA1c for Abbott Systems is assay quality control material consisting of five levels.
Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Frequently Bought Together