Linearity BNP, AUDIT Microcontrols
Supplier: Audit Microcontrols
Certificates
About this item
Universal linearity BNP High, low and extended and linearity lipids for Siemens Centaur, linearity BNP for Siemens Stratus are assayed quality control materials consisting of five levels of human plasma.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Frequently Bought Together