BD BBL™ SIM Prepared Tube Media, BD Diagnostics
Catalog # 90001-180
Supplier: BD
CAS Number:
Specifications
- Pack type:Tube
- Description:SIM media
- Size:10/pack
- Cat. No.:90001-180
- Supplier no.:221010
Specifications
About this item
The SIM prepared tube media is used to differentiate enteric bacilli on the basis of sulfide production, indole formation and motility.
Tube size K.