BBL™ Fluid Thioglycollate Medium, Enriched, BD Diagnostics
Catalog # 90004-930
Supplier: BD
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Fluid thioglycollate medium enriched are general-purpose media used in qualitative procedures for the cultivation of fastidious, as well as nonfastidious microorganisms.
- Used in the isolation and cultivation slow growing obligately anaerobic microorganismls
Media supplemented with sodium bicarbonate or a marble chip is used to prepare a standardized inoculum by the growth method for antimicrobial susceptibility testing of anaerobic bacteria.
Fluid thioglycollate medium, enriched is BBL™ fluid thioglycollate medium supplemented with vitamin K and hemin.
Specifications
Specifications
- Pack type:Carton
- Description:BD BBL™ fluid thioglycollate medium enriched
- Size:100/pack