ContactUsLinkComponent
Glycogen Phosphorylase Colorimetric Assay Kit, BioVision
Glycogen Phosphorylase Colorimetric Assay Kit, BioVision
  76375-826
 :  Avantor
 :  
Glycogen Phosphorylase Colorimetric Assay Kit, BioVision
  76375-826
 :  Avantor
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Glycogen phosphorylase assay kit
  • Cat. No.:
    76375-826
  • No. of assays/kit:
    100 assays
Good for detecting glycogen phosphorylase activity.
  • Sensitive and Rapid
  • Convenient tool for detecting glycogen phosphorylase activity
  • The kit can detect as low as 10 mU in a variety of sample types
Glycogen phosphorylase (EC 2.4.1.1) catalyzes the rate-limiting step in glycogenolysis using glycogen and inorganic phosphate to produce glucose-1-phosphate (G1P). In mammals, glycogen phosphorylase is abundant in muscle, liver, and brain tissues. There are two forms of glycogen phosphorylase, namely glycogen phosphorylase a and b forms.
Glycogen phosphorylase a is the highly active form whereas glycogen phosphorylase b has only limited activity. Glycogen phosphorylase is clinically significant enzyme as its mutations are associated with different glycogen storage diseases in muscle and liver. In addition, this enzyme has been suggested as a biomarker for gastric cancer and its inhibition has been tested in treating type 2 diabetes.
BioVision’s Glycogen Phosphorylase Activity Assay Kit is based on the detection of G1P by a set of enzymatic reactions to generate a colored product with a strong absorbance at 450 nm. The OD 450 nm signal is directly proportional to the glycogen phosphorylase activity. BioVision’s Glycogen Phosphorylase Colorimetric Assay Kit is rapid, sensitive and convenient tool for detecting glycogen phosphorylase activity. The kit can detect as low as 10 µ in a variety of sample types.