BD BBL™ Brain Heart CC Agar with Sheep Blood, Prepared Media Stacker™ Plates, BD Diagnostics
Catalog # 90008-410
Supplier: BD
CAS Number:
Specifications
- Description:Brain heart CC agar with sheep blood (deep fill)
- Size:20 shelfpack
- Cat. No.:90008-410
- Shelf life:112 days
- Supplier no.:296178
Specifications
About this item
Brain heart infusion agar with chloramphenicol, cycloheximide and sheep blood is a selective medium used for the isolation of pathogenic fungi from specimens heavily contaminated with bacteria and saprophytic fungi.
- Ready for immediate use
- Serves as the base for enriched and more selective media supplemented with sheep blood and antibiotics.
With 10% sheep blood, it is used to isolate systemic fungi that may grow poorly on the nonenriched medium. The presence of the antimicrobial agents, cycloheximide and/or chloramphenicol and, in modified formulations, gentamicin, penicillin and streptomycin, inhibits the growth of a wide variety of bacteria and fungi and enhances the isolation of pathogenic fungal species.