Order Entry
United States
ContactUsLinkComponent
Tryptic Soy Broth with 0.5% Polysorbate 80, BD Diagnostics
Tryptic Soy Broth with 0.5% Polysorbate 80, BD Diagnostics
Catalog # 90009-035
Supplier:  BD
CAS Number:  
Tryptic Soy Broth with 0.5% Polysorbate 80, BD Diagnostics
Catalog # 90009-035
Supplier:  BD
Supplier Number:  257366
CAS Number:  

Specifications

  • Pack type:
    Bottle, infusion type
  • Description:
    Tryptic soy broth with 0.5% polysorbate 80
  • Cat. No.:
    90009-035
  • Fill volume:
    100 ml
  • Volume:
    125 ml
  • Supplier no.:
    257366

Specifications

About this item

Tryptic soy broth with polysorbate 80 media are used for sterility testing of specimens containing oils or lecithin.

  • General purpose medium
  • Single wrapped, septum