Order Entry
United States
ContactUsLinkComponent
BD™ Fluid Thioglycollate Medium Bottled Media with 0.5% P80, BD Diagnostics
BD™ Fluid Thioglycollate Medium Bottled Media with 0.5% P80, BD Diagnostics
Catalog # 90009-033
Supplier:  BD
CAS Number:  
BD™ Fluid Thioglycollate Medium Bottled Media with 0.5% P80, BD Diagnostics
Catalog # 90009-033
Supplier:  BD
Supplier Number:  257365
CAS Number:  

Specifications

  • Description:
    BD™ fluid thioglycollate medium with 0.5% P80
  • Size:
    100 ml
  • Cat. No.:
    90009-033
  • Cap type:
    Flip tear-off
  • Supplier no.:
    257365

Specifications

About this item

Fluid thioglycollate medium with 0.5% polysorbate 80 is used for testing oils or materials containing lecithin in sterile fill rooms.

Packaging: Packaged in two Stericlin® bags before autoclaving, providing a sterile outer surface of the containers.
Caution: For laboratory use only.