Linearity Cardiac Markers, Audit MicroControls
Supplier: Audit Microcontrols
Certificates
About this item
Universal Linearity Cardiac Markers, Linearity Cardiac Markers for RAMP, Linearity Cardiac Markers for Siemens Centaur, Linearity Cardiac Markers for Siemens Stratus are assayed quality control materials consisting of five levels of human serum.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Frequently Bought Together