

Potassium hydroxide 0.1 mol/l (0.1 N) in ethanol, Titripur® volumetric solution, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Density:0.836 g/cm³ (20 °C)
Flash Pt:12 °C (EtOH)
EINECS:215-181-3
UN:2924
ADR:3 (8),II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Form:liquid
- Amount-of-substance concentration:0.0995-0.1005
- Measurement uncertainty:+/- 0.0003
- Traceability:NIST SRM
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_1.09115
SDS
MilliporeSigma Supelco® Titration Goes Digital
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together