
Potassium hydroxide 0.5 mol/l (0.5 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Catalog # EM1.11586.5000
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:5 L
- Cat. No.:EM1.11586.5000
- Supplier no.:1.11586.5000
Specifications
About this item
:
:56.11 g/mol
ADR:8,
:M-Clarity™ quality level = MQ100
EINECS:215-181-3
UN:1814
:GHS05
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553