Order Entry
United States
ContactUsLinkComponent
FastCellular™ Senescence Detection Kit, MP Biomedicals
FastCellular™ Senescence Detection Kit, MP Biomedicals
Catalog # 76337-152
Supplier:  MP Biomedicals
CAS Number:  
FastCellular™ Senescence Detection Kit, MP Biomedicals
Catalog # 76337-152
Supplier:  MP Biomedicals
Supplier Number:  092690301
CAS Number:  

Specifications

  • Description:
    Senescence detection kit
  • Size:
    1 Kit
  • Cat. No.:
    76337-152

Specifications

About this item

FastCellular™ senescence detection kit – SPiDER-β-Gal utilizes an improved reagent and method over X-gal for the detection of cellular senescence.

  • Store at 0 to 5 °C with protection from light

The new fluorogenic detection probe offers higher sensitivity and short staining times. The kit is applicable for both living and fixed cells.