Order Entry
United States
ContactUsLinkComponent
FastGro™ FBS Replacement, MP Biomedicals
FastGro™ FBS Replacement, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
FastGro™ FBS Replacement, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • Sterility:
    Sterile
  • Solubility:
    Soluble in water
  • Storage and Stability:
    2...8 °C
  • pH:
    7 - 8
  • Appearance:
    Light opalescent liquid

Specifications

Frequently Bought Together