About this item
pZerve™ Cyropreservation solution is a ready-to-use, sterile, cryopreservation solution lacking dimethyl sulfoxide (DMSO), fetal bovine serum or other animal proteins.
This solution has been successfully used with the following cell types including, but not limited to mast cells, dendritic cells, mesenchymal cells, LNCaP - Metastatic Prostate Adenocarcinoma, Human, LS 180 - Colon, Adenocarcinoma, Human and 293 - Transformed Primary Embryonal Kidney, Human. For use as cell culture and in cell cryopreservation.
This solution does not require any dilution or further processing and can be used for cells cultured in media with and without serum.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Sterility:Sterile
- Storage and Stability:2...8 °C