NorthFlex™ Light Task Plus 5 Polyurethane Palm Coated Dyneema® Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
Knitted glove in Dyneema®, polyamide, and composite fiber with a grey PU coating on the palm. Ideal for handling sharp parts in a dry or greasy environment.
- Conforms snugly to the hand, with good dexterity
- Cool Dyneema® touch provides added comfort
- Tight knit (gauge 13) provides added support, and limits the penetration of dirt
- Ventilated back optimizes breathability
Cut, puncture, and abrasion resistant gloves are ideally suited for use in automotive, manufacturing, construction, steel, mining, paper, and transportation applications and industries.
Specifications
- Catalog No:
- 89092-752
- 89092-756
- 89092-758
- 89092-760
- 89092-754
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Disposable/Reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Finish:
- Palm Coating
- Palm Coating
- Palm Coating
- Palm Coating
- Palm Coating
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff Style:
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile