NorthFlex™ Light Task Plus II™ Polyurethane Palm Coated Dyneema® Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
Gloves feature a Dyneema® seamless liner with white polyurethane coating and knit wrists.
- Dynemma® cut-resistant fiber shell
- White with white polyurethane palm and fingertip coating
- Gloves provide a good cut and abrasion protection level
Lightweight gloves are ideally suited for use in construction, manufacturing, municipal services, and steel and metals applications and industries.
Certifications: Meets ANSI/ISEA 105 - ASTM D3389-05, ANSI/ISEA 105 - ASTM F1790-97, EN 388, and EN 420 requirements.
Packaging: Gloves are packaged 12 pair per pack, 72 pair per case.
Specifications
- Catalog No:
- 89219-016
- 89219-018
- 89219-008
- 89219-010
- 89219-014
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Disposable/Reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Finish:
- Palm and Finger Coating
- Palm and Finger Coating
- Palm and Finger Coating
- Palm and Finger Coating
- Palm and Finger Coating
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff Style:
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- EN Norm:
- EN 388; EN 420
- EN 388; EN 420
- EN 388; EN 420
- EN 388; EN 420
- EN 388; EN 420
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile