NorthFlex™ Red™ Nylon/PVC Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
Lightweight, 15 gauge nylon glove with foam PVC coating on palm and fingertips.
- Nylon glove with foam PVC coating on palm and fingertips
- No coating on back of hand for added ventilation
- Red nylon and black PVC
Lightweight gloves are ideally suited for use in agriculture, construction, manufacturing, municipal services, steel and metal, and utility applications and industries.
Certifications: Meets ANSI/ISEA 105 - ASTM D3389-05, ANSI/ISEA 105 - ASTM F1790-97, EN 388, and EN 420 requirements.
Specifications
- Catalog No:
- 89041-796
- 89041-800
- 89041-802
- 89041-792
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Disposable/Reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Finish:
- Palm and Finger Coating
- Palm and Finger Coating
- Palm and Finger Coating
- Palm and Finger Coating
- Gauge:
- 15
- 15
- 15
- 15
- Cuff Style:
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- EN Norm:
- EN 388; EN 420
- EN 388; EN 420
- EN 388; EN 420
- EN 388; EN 420
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile