Specifications
- Calibration:Up to 4 points
- Temperature Range:0...100 °C (32...212 °F)
- Description:Oakton CON 550 Benchtop Conductivity Meter Kit
- Model:Benchtop
- Temperature Accuracy:±0.5 °C ±1 digit
- Temperature Resolution:0.1/1 °F (0.1 °C)
- Automatic Buffer Recognition:NIST and USA
- Conductivity Accuracy:±1.0% full-scale ±1 digit
- Conductivity Range:0 µS/cm to 200 mS/cm
- Conductivity Resolution:0.1/1 µS/cm; 0.01/0.1 mS/cm
- Data Logging Capacity:50 groups of data
- Display:LCD
- TDS Range:0 - 100 g/l
- TDS Resolution:0.1/1 mg/l, 0.01/0.1 gl
- Temperature Compensation:Automatic or manual from 32...122 °F (0...50 °C)
- Cat. No.:76338-518
Specifications
About this item
Economical meter for benchtop conductivity measurement.
- Automatically recognizes calibration solutions
- Compact design saves valuable benchtop space
- Data logging allows you to store and recall up to 50 groups of data
- IP54-rated housing is dust- and spill-proof
The CON 550 benchtop conductivity meter features a microprocessor chip which provides automatic calibration, automatic temperature compensation, data storage, parameter setup, and Max/Min reading display. The meter reads 0 to 200 mS/cm across four ranges and the four-point conductivity calibration, with automatic solution recognition, makes for easy calibration. The advanced digital processing technology improves meter response time and measuring accuracy. A stable reading display icon appears when the measuring value is stable.
A conductivity electrode with temperature sensor measures conductivity and temperature simultaneously and allows for automatic temperature compensation. The compact meter includes a detachable electrode holder and three bottles of conductivity calibration solution. It has a dustproof and spillproof rating of IP54. Connectors are protected by silicone seal caps.