ICP Calibration Standard 15, (10X) VeriSpec®
15-component standard in 5% nitric acid
50 ppm: Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se Sr, Zn; 500 ppm K
: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Frequently Bought Together