ICP Calibration Standard 19, VeriSpec®
Catalog # RCRV010709100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Matrix:5% HNO₃
- Pack type:Plastic bottle
- Description:VeriSpec® ICP Calibration Standard 19
- Size:100 ml
- Cat. No.:RCRV010709100N
- Supplier no.:RV010709-100N
Specifications
About this item
19-component standard in 5% nitric acid
400 ppm Tl; 200 ppm Bi, In, Pb; 150 ppm Ga, 100 ppm Al; 50 ppm Ag, Ni; 25 ppm Cr; 20 ppm Cd, Co, Cu, Zn; 15 ppm B, Fe; 5 ppm Ba, Mn; 1 ppm Be, Sr
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility