About this item
Thermo Scientific InSight Gen 3 is a remote monitoring and asset management solution that monitors key reliability parameters and uses advanced analytics to provide predictive indicators of possible system failures. InSight consists of Wi-Fi modules (motes), sensors, and software tools that allow you to monitor your laboratory equipment for key performance and reliability parameters and analyze data.
- Asset Predictive Notification allowing you to fix issues before failures occur
- Monitor and track cabinet conditions, ambient conditions, energy consumption/compressor cycles, and door openings from a single device
- Direct to cloud platform connection (no on premise gateway server required)
- New data port only monitoring for compatible devices: Forma 3110, Infors ATR, HeraCell 150/240, HeraCell 160i/250i
InSight provides predictive system health indicators and asset management tools, such as reliability monitoring and repair effectiveness management, energy consumption and asset financial performance, overall asset performance, and health scoping. Alarms are issued automatically to users’ email and mobile devices when threshold breaches are detected in the system. InSight Analytics InSight Analytics, powered by KLATU™, is a system designed to collect, analyze, and make efficiency and operating cost recommendations from multiple asset inputs and events such as: lab equipment health assessment, energy consumption, door openings, and environmental conditions (ambient temperature).
InSight Analytics monitors your compatible laboratory equipment 24 hours a day, seven days a week and alerts users when thresholds are exceeded. The data from InSight Analytics is run through a number of algorithms utilizing the InSight software. With InSight Analytics, you can determine and compare the asset performance against other similar laboratory equipment and product specifications.
How InSight Analytics works: Data is sent wirelessly from the InSight mote via your Wi-Fi network to InSight Cloud Platform. Once in the Cloud, the data is analyzed by the system, providing real-time comparison of the laboratory equipment’s health to industry averages. This data analysis enables you to make knowledge based asset decisions based on key performance indicators such as product service and support, service vs. buy decisions, and remote system health monitoring and trouble-shooting for service providers.
InSight asset management helps ensure the safety and integrity of the lab’s most precious samples by capturing and analyzing performance data. Expert set-up and quality maintenance is key in optimizing the functionality of the system. Thermo Scientific Unity Lab Services offers a full suite of installation, training, system maintenance, and calibration services – all to ensure ideal performance from your InSight monitoring system.